Learn More
Sigma Aldrich Fine Chemicals Biosciences Epigallocatechin 3 galla50MG
Catalog No. 502902637
Encompass
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG
Catalog No. 50-290-2637 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHL8965650MG
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG