Learn More
Medchemexpress LLC (-)-Epigallocatechin-3-O-(4-O-methyl)gallate | 224434-07-5 | MFCD06799453 | 472.40 | 1 MG
SDP
Catalog No. 5000365176
Encompass_Preferred
(-)-Epigallocatechin-3-O-(4-O-methyl)gallate is an orally active catechin derivative found in Taiwanese oolong tea. It can inhibit histamine release and exhibits anti-inflammation and anti-allergic effect. It can be used for researches of immunology and inflammation, such as type I allergic.
  • Inhibits histamine release from IgE/antigen-activated mouse mast cells (45% inhibition rate at 50 μg/mL).
  • Exhibits excellent plasma stability (only 10% degradation rate at 500 μg/mL for 15 mins).
  • Significantly inhibits type I allergic reactions in mice sensitized with ovalbumin (5-50 mg/kg, i.g.).
  • Achieved approximately 50% inhibition rate of abdominal wall blue dye permeation at 10 mg/kg in animal models.
Catalog No. 50-003-65176 Supplier Medchemexpress LLC Supplier No. 1MGHYN15663
(-)-Epigallocatechin-3-O-(4-O-methyl)gallate is an orally active catechin derivative found in Taiwanese oolong tea. It can inhibit histamine release and exhibits anti-inflammation and anti-allergic effect. It can be used for researches of immunology and inflammation, such as type I allergic.
  • Inhibits histamine release from IgE/antigen-activated mouse mast cells (45% inhibition rate at 50 μg/mL).
  • Exhibits excellent plasma stability (only 10% degradation rate at 500 μg/mL for 15 mins).
  • Significantly inhibits type I allergic reactions in mice sensitized with ovalbumin (5-50 mg/kg, i.g.).
  • Achieved approximately 50% inhibition rate of abdominal wall blue dye permeation at 10 mg/kg in animal models.