Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (-)-Epigallocatechin, 95%, from green tea

Catalog No. ac448980100
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448980100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC448980100 Supplier Thermo Scientific Chemicals Supplier No. 448980100
Only null left
Add to Cart

CAS: 970-74-1 | C15H14O7 | 306.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 970-74-1
Molecular Formula C15H14O7
Molecular Weight (g/mol) 306.27
InChI Key XMOCLSLCDHWDHP-IUODEOHRSA-N
Synonym --epigallocatechin, epigallocatechin, epigallocatechol, l-epigallocatechin, epi-gallocatechin, antiscurvy factor c2, --epigallocatechol, l-epigallocatechol, --3,3',4',5,5',7-flavanhexol, 3,3',4',5,5',7-flavanhexol
PubChem CID 72277
ChEBI CHEBI:42255
IUPAC Name (2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)O

Specifications

Assay Percent Range 94% min. (HPLC)
Quantity 10 mg
Formula Weight 306.27
Percent Purity 95%
Chemical Name or Material (-)-Epigallocatechin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.