Learn More
Apexbio Technology LLC Epigallocatechin 970-74-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092570
Encompass_Preferred
Epigallocatechin (CAS 970-74-1) is a naturally occurring polyphenolic compound isolated from green tea It is known to modulate multiple cellular pathways including oxidative stress response and signal transduction primarily through its antioxidant properties and its ability to influence enzyme activities and cellular signaling cascades Soluble in methanol and ethanol epigallocatechin can be stored under cool dry and airtight conditions This compound is widely utilized in biomedical research for investigating its potential roles in cellular protection cancer biology metabolic regulation and as a tool for studying redox-mediated cellular processes
Catalog No. 50-000-92570 Supplier Apexbio Technology LLC Supplier No. N18725.1
Epigallocatechin (CAS 970-74-1) is a naturally occurring polyphenolic compound isolated from green tea It is known to modulate multiple cellular pathways including oxidative stress response and signal transduction primarily through its antioxidant properties and its ability to influence enzyme activities and cellular signaling cascades Soluble in methanol and ethanol epigallocatechin can be stored under cool dry and airtight conditions This compound is widely utilized in biomedical research for investigating its potential roles in cellular protection cancer biology metabolic regulation and as a tool for studying redox-mediated cellular processes