Learn More
Medchemexpress LLC (-)-Epigallocatechin | 970-74-1 | 99.7% | 306.27 | 25 MG
SDP
Catalog No. 5000430668
Encompass_Preferred
(-)-Epigallocatechin, a major flavonoid in green tea, prevents amyloid fibril formation by binding to unfolded polypeptides and stabilizing their native-like state. It also exhibits antimicrobial effects, eradicating E. faecalis biofilms and reducing virulence through hydroxyl radical induction.
  • Most abundant flavonoid in green tea.
  • Binds to unfolded native polypeptides.
  • Prevents amyloid fibril conversion.
  • Stabilizes native-like molecular states.
  • Eradicates E. faecalis biofilms.
  • Induces hydroxyl radicals in E. faecalis.
  • Down-regulates E. faecalis virulence genes.
Catalog No. 50-004-30668 Supplier Medchemexpress LLC Supplier No. 25MGHYN0225
(-)-Epigallocatechin, a major flavonoid in green tea, prevents amyloid fibril formation by binding to unfolded polypeptides and stabilizing their native-like state. It also exhibits antimicrobial effects, eradicating E. faecalis biofilms and reducing virulence through hydroxyl radical induction.
  • Most abundant flavonoid in green tea.
  • Binds to unfolded native polypeptides.
  • Prevents amyloid fibril conversion.
  • Stabilizes native-like molecular states.
  • Eradicates E. faecalis biofilms.
  • Induces hydroxyl radicals in E. faecalis.
  • Down-regulates E. faecalis virulence genes.