Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Epigallocatechin | 970-74-1 | MFCD00075939 | >95.0% | 306.27 | C15H14O7 | 10 MG
SDP

Catalog No. 5000295830
Encompass_Preferred

(-)-Epigallocatechin is an analytical reference standard of a flavonoid commonly found in green tea, provided for research and analytical applications such as studies of matrix metalloproteinases and autophagy. It has molecular formula C15H14O7 and molecular weight 306.27 g/mol. Store and handle according to the Certificate of Analysis; for research use only.

  • Analytical reference standard for method development and quality control.
  • Intended for research and analytical applications, including MMP and autophagy assays.
  • Molecular formula C15H14O7 and molecular weight 306.27 g/mol.
  • Purity >95.0% (HPLC).
  • Supplied in milligram-scale quantities for analytical use; follow CoA for storage and handling.

Catalog No. 50-002-95830 Supplier Medchemexpress LLC Supplier No. HYN0225R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Epigallocatechin is an analytical reference standard of a flavonoid commonly found in green tea, provided for research and analytical applications such as studies of matrix metalloproteinases and autophagy. It has molecular formula C15H14O7 and molecular weight 306.27 g/mol. Store and handle according to the Certificate of Analysis; for research use only.

  • Analytical reference standard for method development and quality control.
  • Intended for research and analytical applications, including MMP and autophagy assays.
  • Molecular formula C15H14O7 and molecular weight 306.27 g/mol.
  • Purity >95.0% (HPLC).
  • Supplied in milligram-scale quantities for analytical use; follow CoA for storage and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.