Learn More
Abcam (-)-Epigallocatechin, Antio x idant activity, 10MG
Catalog No. 502706259
Encompass_Preferred
MW 306.27 Da, Purity >98%. Antioxidant and anti-tumor promoter activity. Induces apoptosis and inhibits angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6259 Supplier Abcam Supplier No. AB14237810MG
MW 306.27 Da, Purity >98%. Antioxidant and anti-tumor promoter activity. Induces apoptosis and inhibits angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement