Learn More
Apexbio Technology LLC (-)-Epigallocatechin gallate (EGCG) 989-51-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063001
Encompass_Preferred
(-)-Epigallocatechin gallate (EGCG CAS 989-51-5) is a major catechin component derived from green tea comprising approximately 59% of total catechins EGCG exhibits antioxidant antiangiogenic antitumor and antiviral properties by modulating cellular signaling pathways involved in apoptosis induction cell cycle arrest and tumorigenesis inhibition It suppresses viral replication in pathogens including HCV HIV-1 HBV HSV-1/2 EBV adenovirus influenza virus and enterovirus and inhibits enzyme targets such as DNA methyltransferases (DNMTs) proteases and DHFR EGCG is utilized widely in biomedical research examining chemoprevention mechanisms and therapeutic potential in cancers such as hepatic gastric dermal pulmonary breast and colorectal malignancies
Catalog No. 50-000-63001 Supplier Apexbio Technology LLC Supplier No. A26005.1
(-)-Epigallocatechin gallate (EGCG CAS 989-51-5) is a major catechin component derived from green tea comprising approximately 59% of total catechins EGCG exhibits antioxidant antiangiogenic antitumor and antiviral properties by modulating cellular signaling pathways involved in apoptosis induction cell cycle arrest and tumorigenesis inhibition It suppresses viral replication in pathogens including HCV HIV-1 HBV HSV-1/2 EBV adenovirus influenza virus and enterovirus and inhibits enzyme targets such as DNA methyltransferases (DNMTs) proteases and DHFR EGCG is utilized widely in biomedical research examining chemoprevention mechanisms and therapeutic potential in cancers such as hepatic gastric dermal pulmonary breast and colorectal malignancies