Learn More
AdipoGen (-)-Epigallocatechin gallate (10 mg)
SDP
Supplier:  AdipoGen CDXE0128M010
Encompass_Preferred
Chemical. CAS: 989-51-5. Formula: C22H18O11. MW: 458.37. (-)-Epigallocatechin gallate (EGCG), an antioxidant and anti-inflammatory polyphenol flavonoid exerts antitumor properties by inhibiting telomerase and DNA methyltransferase activity. EGCG inhibits the expression of matrix metalloproteinase-2 (MMP-2), MMP-9 and reduces the invasiveness. EGCG blocks the activation of epidermal growth factor (EGF) receptors and human epidermal growth factor receptor-2 (HER-2). Regulates cancer cell growth, proliferation, transformation, survival, angiogenesis, apoptosis, invasion and metastasis. EGCG increases bone mineral density and reduces bone resorption. EGCG inhibits osteoclastogenesis by inhibiting receptor activator of nuclear factor kappaB ligand (RANKL) induced nuclear factor kappa B (NF-kappaB) transcriptional activity. EGCG has neuroprotective and anti-aging property and increases myogenic differentiation. EGCG inhibits fatty acid synthase and glutamate dehydrogenase activity.
Catalog No. 50-205-4414