Learn More
AdipoGen (-)-Epigallocatechin gallate (25 mg)
SDP
Catalog No. 502053410
Encompass_Preferred
Chemical. CAS: 989-51-5. Formula: C22H18O11. MW: 458.4. Isolated from green tea. Potent anticancer compound. Anti-angiogenic. VEGF, VE-cadherin phosphorylation, matrix metalloproteinase and urokinase-plasminogen activator (uPA) inhibitor. Anti-inflammatory. NF-kappaB inhibitor. Modulates chronic inflammatory diseases, such as type 2 diabetes, rheumatoid arthritis, inflammatory bowel disease. COX-2 and inducible nitric oxide synthase (iNOS; NOS II) inhibitor. Potent antioxidant. Protects cells from lipid peroxidation and DNA damage induced by reactive free radicals. Regulates cancer cell growth, proliferation, transformation, survival, angiogenesis, apoptosis, invasion and metastasis. Chemopreventive. Apoptosis inducer. Promotes cell cycle arrest. Modulates signal transduction pathways including JAK/STAT, MAPK, PI3K/AKT, Wnt and Notch. EGFR and HER-2 receptor signaling inhibitor. MAPKs and activator protein-1 inhibitor. mTOR suppressor.
Catalog No. 50-205-3410 Supplier AdipoGen Supplier No. AGCN20063M025
Chemical. CAS: 989-51-5. Formula: C22H18O11. MW: 458.4. Isolated from green tea. Potent anticancer compound. Anti-angiogenic. VEGF, VE-cadherin phosphorylation, matrix metalloproteinase and urokinase-plasminogen activator (uPA) inhibitor. Anti-inflammatory. NF-kappaB inhibitor. Modulates chronic inflammatory diseases, such as type 2 diabetes, rheumatoid arthritis, inflammatory bowel disease. COX-2 and inducible nitric oxide synthase (iNOS; NOS II) inhibitor. Potent antioxidant. Protects cells from lipid peroxidation and DNA damage induced by reactive free radicals. Regulates cancer cell growth, proliferation, transformation, survival, angiogenesis, apoptosis, invasion and metastasis. Chemopreventive. Apoptosis inducer. Promotes cell cycle arrest. Modulates signal transduction pathways including JAK/STAT, MAPK, PI3K/AKT, Wnt and Notch. EGFR and HER-2 receptor signaling inhibitor. MAPKs and activator protein-1 inhibitor. mTOR suppressor.