Learn More
STEMCELL Technologies (-)-Epigallocatechin Gallate, Size: 100 mg
SDP
Supplier:  STEMCELL Technologies 73644
Encompass
Inhibit DNA methyltransferases (Lee et al. Mol Pharmacol, 2005) and the formation of oxidized low-density lipoproteins (Yoshida et al. Biochem Pharmacol, 1999) with (-)-Epigallocatechin Gallate. This polyphenol catechin antioxidant inhibits bone resorption by inducing cell death of osteoclast-like multinucleated cells (Nakagawa et al. Biochem Biophys Res Commun, 2002); inhibits growth and induces apoptosis in human pancreatic cancer cells in mouse xenograft model (Shankar et al. Front Biosci, 2008, Du et al. Nutrients, 2012); and causes cell cycle deregulation and apoptosis in human epidermoid cancer cell line, possibly via inhibition of NF-κB (Ahmad et al. Arch Biochem Biophys, 2000). (-)-Epigallocatechin Gallate is supplied as a crystalline solid with ≥98% purity; Molecular weight 458.4 g/mol; CAS Number 989-51-5. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-6107