Learn More
TARGETMOL CHEMICALS INC Epigallocatechin Gallate 500MG
SDP
Catalog No. 502877643
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (-)-Epigallocatechin Gallate (EGCG) is a phenolic antioxidant polyphenol flavonoid found in plants such as green and black tea, which inhibits telomerase and DNA methyltransferase, blocks the activation of EGF receptors and HER-2 receptors, inhibits cellular oxidation, and prevents free radical damage to cells. Purity 99.34%
Catalog No. 50-287-7643 Supplier TARGETMOL CHEMICALS INC Supplier No. T2988500MG
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (-)-Epigallocatechin Gallate (EGCG) is a phenolic antioxidant polyphenol flavonoid found in plants such as green and black tea, which inhibits telomerase and DNA methyltransferase, blocks the activation of EGF receptors and HER-2 receptors, inhibits cellular oxidation, and prevents free radical damage to cells. Purity 99.34%