Learn More
Medchemexpress LLC (-)-Epigallocatechin Gallate | 989-51-5 | >98% | 458.37 | 1 G
SDP
Catalog No. 5000352295
Encompass_Preferred
(-)-Epigallocatechin Gallate (EGCG) is a major polyphenol found in green tea. It inhibits cell proliferation and induces cell apoptosis, and inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity. It possesses potent anticancer, antioxidant, and anti-inflammatory properties against various types of cancers, including colorectal cancer, myeloid leukemia, and thyroid carcinoma.
  • Inhibits cell proliferation
  • Induces cell apoptosis
  • Inhibits glutamate dehydrogenase 1/2 activity
  • Possesses anticancer, antioxidant, and anti-inflammatory properties
  • Effective against various cancers such as colorectal cancer, myeloid leukemia, and thyroid carcinoma
Catalog No. 50-003-52295 Supplier Medchemexpress LLC Supplier No. HY136531G
(-)-Epigallocatechin Gallate (EGCG) is a major polyphenol found in green tea. It inhibits cell proliferation and induces cell apoptosis, and inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity. It possesses potent anticancer, antioxidant, and anti-inflammatory properties against various types of cancers, including colorectal cancer, myeloid leukemia, and thyroid carcinoma.
  • Inhibits cell proliferation
  • Induces cell apoptosis
  • Inhibits glutamate dehydrogenase 1/2 activity
  • Possesses anticancer, antioxidant, and anti-inflammatory properties
  • Effective against various cancers such as colorectal cancer, myeloid leukemia, and thyroid carcinoma