Learn More
Medchemexpress LLC (-)-epigallocatechin gallate | 989-51-5 | MFCD00075940 | 98.1% | C22H18O11 | 10MG
SDP
Catalog No. 5000259369
Encompass_Preferred
(-)-Epigallocatechin gallate (EGCG) is a flavonoid phenol supplied as a high-purity analytical reference standard for chromatography and mass spectrometry. It is provided as a solid for research applications; users should follow the certificate of analysis for storage and handling.
  • Analytical standard for HPLC, GC, and MS method development and quantitation.
  • High purity: 98.1% (rounded from COA).
  • Chemical formula C22H18O11; molecular weight 458.37 g/mol.
  • Available in small package sizes suitable for analytical assays.
  • Physical appearance: white to off-white solid.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-59369 Supplier Medchemexpress LLC Supplier No. HY13653R10MG
(-)-Epigallocatechin gallate (EGCG) is a flavonoid phenol supplied as a high-purity analytical reference standard for chromatography and mass spectrometry. It is provided as a solid for research applications; users should follow the certificate of analysis for storage and handling.
  • Analytical standard for HPLC, GC, and MS method development and quantitation.
  • High purity: 98.1% (rounded from COA).
  • Chemical formula C22H18O11; molecular weight 458.37 g/mol.
  • Available in small package sizes suitable for analytical assays.
  • Physical appearance: white to off-white solid.
  • Intended for research use only; not for human or clinical use.