Learn More
Abcam (-)-Epigallocatechin gallate, antio x idant flavonoid, 500MG
Catalog No. 502707342
Encompass_Preferred
MW 458.38 g/mol, Purity >97%. Potent antioxidant flavonoid. Displays anticancer, antitumour activity and induces apoptosis. Modulates numerous cell signalling pathways, such as inhibiting NF-κB, MAPK and COX-2 pathways. Also shows anti-inflammatory, antibacterial and antiviral activities (EC₅₀ = ~22 μM for inhibition of influenza virus).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7342 Supplier Abcam Supplier No. AB120716500MG
MW 458.38 g/mol, Purity >97%. Potent antioxidant flavonoid. Displays anticancer, antitumour activity and induces apoptosis. Modulates numerous cell signalling pathways, such as inhibiting NF-κB, MAPK and COX-2 pathways. Also shows anti-inflammatory, antibacterial and antiviral activities (EC₅₀ = ~22 μM for inhibition of influenza virus).
The product is subject to the following: Abcam Restricted Use Statement