Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Epigallocatechin Gallate (EGCG) | 989-51-5 | 99.75% | 458.37 | 10 MM * 1 ML
SDP

Catalog No. 5000348439
Encompass_Preferred

(-)-Epigallocatechin Gallate (EGCG) is a major polyphenol found in green tea. It can inhibit cell proliferation and induce cell apoptosis, and it inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity. EGCG possesses potent anticancer, antioxidant, and anti-inflammatory properties against various types of cancers such as colorectal cancer, myeloid leukemia, and thyroid carcinoma.

  • Can inhibit cell proliferation and induce cell apoptosis.
  • Inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity.
  • Potent anticancer, antioxidant, and anti-inflammatory properties against various types of cancers such as colorectal cancer, myeloid leukemia, thyroid carcinoma.

Catalog No. 50-003-48439 Supplier Medchemexpress LLC Supplier No. HY1365310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Epigallocatechin Gallate (EGCG) is a major polyphenol found in green tea. It can inhibit cell proliferation and induce cell apoptosis, and it inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity. EGCG possesses potent anticancer, antioxidant, and anti-inflammatory properties against various types of cancers such as colorectal cancer, myeloid leukemia, and thyroid carcinoma.

  • Can inhibit cell proliferation and induce cell apoptosis.
  • Inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity.
  • Potent anticancer, antioxidant, and anti-inflammatory properties against various types of cancers such as colorectal cancer, myeloid leukemia, thyroid carcinoma.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.