Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epigallocatechin gallate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111016011 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6011 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6011 Supplier MilliporeSigma Supelco Supplier No. PHR13331G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Epigallocatechin gallate is a potent polyphenolic flavonoid which is found as a component of tea. It exhibits antioxidant, antimutagenic, antitumor and anti-inflammatory properties, thereby contributing to the health-beneficial actions. It finds potential use as a drug candidate in the pharmaceutical, cosmetic, and nutritional fields. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 989-51-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C22H18O11
MDL Number MFCD00075940
Synonym (-)-Epigallocatechin gallate; (-)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2 H)-benzopyran-3,5,7-triol 3-gallate; (-)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate; EGCG
Type Certified Reference Material
Quantity 1 g
Formula Weight 458.37
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.