Learn More
(-)-Epigallocatechin Gallate, Spectrum™ Chemical
SDP
Catalog No. 18620689 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
50 mg
Catalog No. Quantity
18-620-689 50 mg
1 options
Catalog No. 18-620-689 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E115450MG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Synonym: Epigallocatechin Gallate, Epigallocatechin 3-gallate
(-)-Epigallocatechin Gallate is an antioxidant polyphenol flavonoid isolated from green tea. Its possible benefit as a nutritional chemopreventive agent for cancer, atherosclerosis, and neurodegenerative diseases is generating increased scientific interest. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Specifications
Quantity 50 mg