Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Epigallocatechin Gallate (Standard) | 989-51-5 | 98.05% | 458.37 | 25 MG
SDP

Catalog No. 5000351926
Encompass_Preferred

(-)-Epigallocatechin Gallate (Standard) is the analytical standard of (-)-Epigallocatechin Gallate, intended for research and analytical applications. It is a major polyphenol in green tea that can inhibit cell proliferation and induce cell apoptosis. It also inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity and possesses potent anticancer, antioxidant, and anti-inflammatory properties against various cancers like colorectal cancer, myeloid leukemia, and thyroid carcinoma.

  • Analytical standard for research and analytical applications
  • Inhibits cell proliferation and induces cell apoptosis
  • Inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity
  • Possesses potent anticancer, antioxidant, and anti-inflammatory properties
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS

Catalog No. 50-003-51926 Supplier Medchemexpress LLC Supplier No. 25MGHY13653R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Epigallocatechin Gallate (Standard) is the analytical standard of (-)-Epigallocatechin Gallate, intended for research and analytical applications. It is a major polyphenol in green tea that can inhibit cell proliferation and induce cell apoptosis. It also inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity and possesses potent anticancer, antioxidant, and anti-inflammatory properties against various cancers like colorectal cancer, myeloid leukemia, and thyroid carcinoma.

  • Analytical standard for research and analytical applications
  • Inhibits cell proliferation and induces cell apoptosis
  • Inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity
  • Possesses potent anticancer, antioxidant, and anti-inflammatory properties
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.