Learn More
Medchemexpress LLC (-)-Epigallocatechin Gallate (Standard) | 989-51-5 | 98.1% | 458.37 | 50 MG
SDP
Catalog No. 5000352300
Encompass_Preferred
(-)-Epigallocatechin Gallate (Standard) is an analytical standard and a major polyphenol found in green tea. It is intended for research and analytical applications, known for its ability to inhibit cell proliferation and induce cell apoptosis. It also inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity and possesses potent anticancer, antioxidant, and anti-inflammatory properties.
  • Analytical standard for (-)-Epigallocatechin Gallate.
  • Intended for research and analytical applications.
  • Potent anticancer, antioxidant, and anti-inflammatory properties against various cancers like colorectal cancer, myeloid leukemia, and thyroid carcinoma.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as a reference standard.
Catalog No. 50-003-52300 Supplier Medchemexpress LLC Supplier No. HY13653R50MG
(-)-Epigallocatechin Gallate (Standard) is an analytical standard and a major polyphenol found in green tea. It is intended for research and analytical applications, known for its ability to inhibit cell proliferation and induce cell apoptosis. It also inhibits glutamate dehydrogenase 1/2 (GDH1/2, GLUD1/2) activity and possesses potent anticancer, antioxidant, and anti-inflammatory properties.
  • Analytical standard for (-)-Epigallocatechin Gallate.
  • Intended for research and analytical applications.
  • Potent anticancer, antioxidant, and anti-inflammatory properties against various cancers like colorectal cancer, myeloid leukemia, and thyroid carcinoma.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as a reference standard.