Learn More
Medchemexpress LLC (-)-Epigallocatechin (Standard) | 970-74-1 | 99.5% | 306.27 | 50 MG
SDP
Catalog No. 5000431049
Encompass_Preferred
(-)-Epigallocatechin (Standard) is the analytical standard of (-)-Epigallocatechin. It is intended for research and analytical applications. It is the most abundant flavonoid found in green tea and has the ability to bind to unfolded native polypeptides, thereby preventing their conversion to amyloid fibrils. As an analytical standard, it is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Most abundant flavonoid found in green tea
  • Binds to unfolded native polypeptides, preventing amyloid fibril conversion
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
Catalog No. 50-004-31049 Supplier Medchemexpress LLC Supplier No. HYN0225R50MG
(-)-Epigallocatechin (Standard) is the analytical standard of (-)-Epigallocatechin. It is intended for research and analytical applications. It is the most abundant flavonoid found in green tea and has the ability to bind to unfolded native polypeptides, thereby preventing their conversion to amyloid fibrils. As an analytical standard, it is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Most abundant flavonoid found in green tea
  • Binds to unfolded native polypeptides, preventing amyloid fibril conversion
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS