Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Epigallocatechin 98.0+%, TCI America™
SDP

Catalog No. p-7129320 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E108410MG 10 mg
E108450MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E108410MG Supplier TCI America Supplier No. E108410MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 970-74-1
Molecular Formula C15H14O7
Molecular Weight (g/mol) 306.27
MDL Number MFCD00075939
InChI Key XMOCLSLCDHWDHP-IUODEOHRSA-N
Synonym --epigallocatechin, epigallocatechin, epigallocatechol, l-epigallocatechin, epi-gallocatechin, antiscurvy factor c2, --epigallocatechol, l-epigallocatechol, --3,3',4',5,5',7-flavanhexol, 3,3',4',5,5',7-flavanhexol
PubChem CID 72277
ChEBI CHEBI:42255
IUPAC Name (2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 10 mg
Formula Weight 306.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Epigallocatechin
RTECSNumber KB5100000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.