Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Epigambogic acid | 887606-04-4 | 1 MG
SDP

Catalog No. 5000419710
Encompass_Preferred

Epigambogic acid is a chemical compound with the molecular formula C38H44O8 and a molecular weight of 628.75. It appears as a yellow to orange solid and has a high purity of 98.63% by HPLC analysis.

  • Molecular formula: C38H44O8
  • Molecular weight: 628.75
  • Appearance: Yellow to orange solid
  • Purity (HPLC): 98.63%
  • Storage: -20°C, sealed storage, away from moisture
  • In solvent storage: -80°C for 6 months, or -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-19710 Supplier Medchemexpress LLC Supplier No. HYN81971MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Epigambogic acid is a chemical compound with the molecular formula C38H44O8 and a molecular weight of 628.75. It appears as a yellow to orange solid and has a high purity of 98.63% by HPLC analysis.

  • Molecular formula: C38H44O8
  • Molecular weight: 628.75
  • Appearance: Yellow to orange solid
  • Purity (HPLC): 98.63%
  • Storage: -20°C, sealed storage, away from moisture
  • In solvent storage: -80°C for 6 months, or -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.