Learn More
Enzo Life Sciences SCREEN-WELL Epigenetics library (100µl/well)
SDP
Supplier:  Enzo Life Sciences BML28360100
Encompass_Preferred
The SCREEN-WELL Epigenetics library contains 43 compounds with defined activity against enzymes which carry out epigenetic modification of lysine. It also includes DNA methylation inhibitors. Compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates at 100 or 500µl per well. The library is a useful tool for chemical genomics, assay development and other pharmacological applications. The library contains inhibitors of these important enzymes: HDACs, SIRTs, Lysine demethylases, HATs, Histone methyl transferases, DNA methyltransferases as well as SIRT activators. A variety of structurally and mechanistically different compound classes are included. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0213