Learn More
Epigentek Inc EpiQuik Chromatin Immunoprecipitation (ChIP) Kit (24 reactions)
SDP
Catalog No. 501096129
Encompass_Preferred
For immunoprecipitating chromatin specifically from cell input samples via microplate format
Catalog No. 50-109-6129 Supplier Epigentek Inc Supplier No. P20021
For immunoprecipitating chromatin specifically from cell input samples via microplate format