Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC EPIKURE 3140
SDP
Catalog No. NC1486450
Encompass_Preferred
EPIKURE Curing Agent 3140 is a low viscosity reactive polyamide, high imidazoline, moderate molecular weight epoxy curing agent based on dimerized fatty acid and polyamines. Epoxy resin compatibility and thin film curves are very good. Benefits include:Good chemical and corrosion resistanceGood water resistanceGood pigment and substrate wettingGood adhesion
Catalog No. NC1486450 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. EPIKURE 3140
EPIKURE Curing Agent 3140 is a low viscosity reactive polyamide, high imidazoline, moderate molecular weight epoxy curing agent based on dimerized fatty acid and polyamines. Epoxy resin compatibility and thin film curves are very good. Benefits include:Good chemical and corrosion resistanceGood water resistanceGood pigment and substrate wettingGood adhesion