Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ EpiLife™ Defined Growth Supplement (EDGS)

Catalog No. S0125
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0125 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0125 Supplier Gibco™ Supplier No. S0125
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

EpiLife™ Defined Growth Supplement (EDGS)

A defined, sterile, concentrated (100X) solution intended for use with EpiLife™ medium to culture human epidermal keratinocytes. EDGS is not intended for use with Medium 154. EDGS contains all of the growth factors and hormones necessary for the growth of human epidermal keratinocytes. Each bottle contains the correct amount of supplement for a 500 ml bottle of EpiLife™ medium. EDGS contains:
  • purified bovine serum albumin
  • purified bovine transferrin
  • hydrocortisone
  • recombinant human insulin-like growth factor type-1 (rhIGF-1)
  • prostaglandin E2 (PGE2)
  • recombinant human epidermal growth factor (rhEGF) The bovine products in EDGS have been isolated from animals of North American origin. The isolation process for these components includes steps that inactivate viruses. All components of EDGS are greater than 95% pure.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage minus 20 degrees C
    Cell Type Keratinocyte Cells
    Concentration 100 X
    Product Type Growth Supplement
    Sterility Sterile
    Product Line Cascade Biologics, EpiLife
    Quantity 5 mL
    Shipping Condition Dry Ice
    Species Human
    Are the Keratinocyte SFM basal medium and supplement kit available separately?

    The basal medium is not available for purchase individually, and can only be ordered as part of the Keratinocyte SFM kit (catalog # 17005-042). The supplements are available separately: Bovine Pituitary Extract is catalog # 13028-014 and the EGF is catalog # PHG0311.

    Which serum-free media can be used to culture mouse keratinocytes?

    The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.

    Which serum-free media can be used to culture human keratinocytes?

    There are a variety of options for serum-free culture of human keratinocytes. Keratinocyte SFM (Cat. No. 17005-042) is a widely used kit that includes the basal medium, bovine pituitary extract (BPE) and Epidermal Growth Factor (EGF). If you would like a serum-free, defined culture system, the options are the Defined Keratinocyte SFM kit (Cat. No. 10744-019) or Epilife medium (Cat. No. M-EPI-500-CA) supplemented with EDGS (Cat. No. S-012-5). If you would like an animal origin free defined system, Epilife medium (Cat. No. M-EPI-500-CA) with S7 supplement (Cat. No. S-017-5) is an excellent choice.

    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.