Learn More
Epigentek Inc EpiMag HT (96-Well) Magnetic Separator (1/Pack)
SDP
Catalog No. NC0684361
Encompass_Preferred
The EpiMag HT (96-Well) Magnetic Separator is a magnetic stand (magnetic rack) that allows paramagnetic bead precipitation of liquid samples from various flat-bottom or U-bottom 96-well microplates (we recommend our EpiMag 96-Well Microplates for ideal compatibility). It can be used for isolation and purification of nucleic acids and proteins, immunoprecipitation, immunoassays (ELISA), cell sorting, and purification of biomolecules. A PCR adapter is also included for the option of using V-bottom semi-skirted/unskirted PCR microplates.
Catalog No. NC0684361 Supplier Epigentek Inc Supplier No. Q100021
The EpiMag HT (96-Well) Magnetic Separator is a magnetic stand (magnetic rack) that allows paramagnetic bead precipitation of liquid samples from various flat-bottom or U-bottom 96-well microplates (we recommend our EpiMag 96-Well Microplates for ideal compatibility). It can be used for isolation and purification of nucleic acids and proteins, immunoprecipitation, immunoassays (ELISA), cell sorting, and purification of biomolecules. A PCR adapter is also included for the option of using V-bottom semi-skirted/unskirted PCR microplates.