Learn More
New England Biolabs, Inc. EpiMark Hot Start Taq DNA Polymerase – 500 reactions
SDP
Supplier:  New England Biolabs, Inc. M0490L
Encompass
EpiMark Hot Start Taq DNA Polymerase is a mixture of Taq DNA Polymerase and a temperature sensitive, aptamer-based inhibitor. The inhibitor binds reversibly to the enzyme, inhibiting polymerase activity at temperatures below 45C, but releases the enzyme during normal PCR cycling conditions. This permits assembly of reactions at room temperature. An advantage of the aptamer-based hot start mechanism is that it does not require a separate high temperature incubation step to activate the enzyme. The advanced aptamer-based hot-start activity coupled with the supplied optimized reaction buffer makes the EpiMark Hot Start Taq DNA Polymerase an excellent choice for use on bisulfite-converted DNA. Taq DNA polymerase possesses a 5- 3 polymerase activity (1,2,3) and a 5 flap endonuclease activity (4,5). EpiMark Hot Start Taq DNA Polymerase is supplied with 5X EpiMark Hot Start Taq Reaction Buffer, providing robust amplification for bisulfite-converted DNA and AT-rich amplicons.
Catalog No. 50-440-1