Learn More
Medchemexpress LLC HY-N0258 5mg Medchemexpress, Epimedin A1 CAS:140147-77-9 Purity:>98%
SDP
Catalog No. 501933496
Encompass_Preferred
Medchemexpress, HY-N0258 5mg Epimedin A1 CAS:140147-77-9 Epimedin A1 is a flavonoid extracted from Herba Epimedii which is one of commonly used Chinese medicines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3496 Supplier Medchemexpress LLC Supplier No. HYN02585MG
Medchemexpress, HY-N0258 5mg Epimedin A1 CAS:140147-77-9 Epimedin A1 is a flavonoid extracted from Herba Epimedii which is one of commonly used Chinese medicines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.