Learn More
Cayman Chemical Epimedin A, Purity: ≥98%, CAS: 110623-72-8, 10 mg
SDP
Catalog No. 502938396
Encompass_Preferred
A flavonoid glycoside; inhibits TPA-induced ear edema in mice (ED50 = 252 nmol/ear); prevents prednisolone-induced osteoporosis in zebrafish
Catalog No. 50-293-8396 Supplier Cayman Chemical Supplier No. 2758210 MG
A flavonoid glycoside; inhibits TPA-induced ear edema in mice (ED50 = 252 nmol/ear); prevents prednisolone-induced osteoporosis in zebrafish