Learn More
Cayman Chemical Epimedin A, Purity: ≥98%, CAS: 110623-72-8, 50 mg
SDP
Catalog No. 502941097
Encompass_Preferred
A flavonoid glycoside; inhibits TPA-induced ear edema in mice (ED50 = 252 nmol/ear); prevents prednisolone-induced osteoporosis in zebrafish
Catalog No. 50-294-1097 Supplier Cayman Chemical Supplier No. 2758250 MG
A flavonoid glycoside; inhibits TPA-induced ear edema in mice (ED50 = 252 nmol/ear); prevents prednisolone-induced osteoporosis in zebrafish