Learn More
Medchemexpress LLC HY-B0640 50mg Medchemexpress, Epinastine CAS:80012-43-7 Purity:>98%
SDP
Catalog No. 502082627
Encompass_Preferred
Medchemexpress, HY-B0640 50mg Epinastine CAS:80012-43-7 Epinastine (WAL801) is an antihistamine and mast cell stabilizer. Epinastine is a potent, selective and orally-active histamine H1 receptor antagonist. Epinastine also inhibits IL-8 release and has an antiallergic action[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2627 Supplier Medchemexpress LLC Supplier No. HYB064050MG
Medchemexpress, HY-B0640 50mg Epinastine CAS:80012-43-7 Epinastine (WAL801) is an antihistamine and mast cell stabilizer. Epinastine is a potent, selective and orally-active histamine H1 receptor antagonist. Epinastine also inhibits IL-8 release and has an antiallergic action[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.