Learn More
Apexbio Technology LLC Epinastine HCl 108929-04-0 50mg
SDP
Catalog No. 502240791
Encompass_Preferred
Epinastine hydrochloride (CAS 108929-04-0) is a small molecule antagonist of the histamine H1 receptor and a mast cell stabilizer By inhibiting H1 receptor-mediated signaling and preventing the degranulation of mast cells epinastine suppresses the release of pro-inflammatory mediators such as histamine It is commonly applied in ophthalmic research models to investigate mechanisms underlying allergic conjunctival inflammation and to evaluate antiallergic therapies Epinastine hydrochloride is used to study the modulation of allergic responses at the cellular and molecular levels particularly within ocular tissues
This item is not returnable. View return policy
Catalog No. 50-224-0791 Supplier Apexbio Technology LLC Supplier No. B174450
Epinastine hydrochloride (CAS 108929-04-0) is a small molecule antagonist of the histamine H1 receptor and a mast cell stabilizer By inhibiting H1 receptor-mediated signaling and preventing the degranulation of mast cells epinastine suppresses the release of pro-inflammatory mediators such as histamine It is commonly applied in ophthalmic research models to investigate mechanisms underlying allergic conjunctival inflammation and to evaluate antiallergic therapies Epinastine hydrochloride is used to study the modulation of allergic responses at the cellular and molecular levels particularly within ocular tissues