Learn More
Medchemexpress LLC Epinastine hydrochloride | 108929-04-0 | 99.86% | 50 MG
SDP
Catalog No. 5000408082
Encompass_Preferred
Epinastine hydrochloride is a selective and orally active histamine H1 receptor antagonist, a CD96/PVR inhibitor, and a mast cell stabilizer. It demonstrates high affinity for neuronal octopamine receptors and inhibits TARC, IL-8, and IL-4. It can activate anti-colon cancer immunity and inhibit Substance P-induced scratching behavior and increased vascular permeability. Epinastine hydrochloride is utilized in the research of allergic diseases.
  • Selective and orally active histamine H1 receptor antagonist
  • CD96/PVR inhibitor
  • Mast cell stabilizer
  • Demonstrates high affinity for neuronal octopamine receptors
  • Inhibits TARC, IL-8, and IL-4
  • Activates anti-colon cancer immunity
  • Inhibits Substance P-induced scratching behavior and increased vascular permeability
  • Utilized in the research of allergic diseases
Catalog No. 50-004-08082 Supplier Medchemexpress LLC Supplier No. HYB0640A50MG
Epinastine hydrochloride is a selective and orally active histamine H1 receptor antagonist, a CD96/PVR inhibitor, and a mast cell stabilizer. It demonstrates high affinity for neuronal octopamine receptors and inhibits TARC, IL-8, and IL-4. It can activate anti-colon cancer immunity and inhibit Substance P-induced scratching behavior and increased vascular permeability. Epinastine hydrochloride is utilized in the research of allergic diseases.
  • Selective and orally active histamine H1 receptor antagonist
  • CD96/PVR inhibitor
  • Mast cell stabilizer
  • Demonstrates high affinity for neuronal octopamine receptors
  • Inhibits TARC, IL-8, and IL-4
  • Activates anti-colon cancer immunity
  • Inhibits Substance P-induced scratching behavior and increased vascular permeability
  • Utilized in the research of allergic diseases