Learn More
Medchemexpress LLC Epinastine hydrochloride | 108929-04-0 | 99.9% | 100 MG
SDP
Catalog No. 5000408229
Encompass_Preferred
Epinastine hydrochloride is a selective and orally active histamine H1 receptor antagonist, CD96/PVR inhibitor, and mast cell stabilizer. It exhibits high affinity for neuronal octapamine receptors in locusts (Ki = 2 nM) and honeybees (Ki = 1.1 nM) and is used in the research of allergic diseases.
  • Selective and orally active histamine H1 receptor antagonist
  • CD96/PVR inhibitor
  • Mast cell stabilizer
  • High affinity for neuronal octapamine receptors
  • Inhibits TARC, IL-8, and IL-4
  • Activates anti-colon cancer immunity
  • Inhibits Substance P-induced scratching behavior and increased vascular permeability
Catalog No. 50-004-08229 Supplier Medchemexpress LLC Supplier No. HYB0640A100MG
Epinastine hydrochloride is a selective and orally active histamine H1 receptor antagonist, CD96/PVR inhibitor, and mast cell stabilizer. It exhibits high affinity for neuronal octapamine receptors in locusts (Ki = 2 nM) and honeybees (Ki = 1.1 nM) and is used in the research of allergic diseases.
  • Selective and orally active histamine H1 receptor antagonist
  • CD96/PVR inhibitor
  • Mast cell stabilizer
  • High affinity for neuronal octapamine receptors
  • Inhibits TARC, IL-8, and IL-4
  • Activates anti-colon cancer immunity
  • Inhibits Substance P-induced scratching behavior and increased vascular permeability