Learn More
Medchemexpress LLC Epinastine hydrochloride | 108929-04-0 | 99.9% | 500 MG
SDP
Catalog No. 5000408738
Encompass_Preferred
Epinastine hydrochloride is a selective and orally active histamine H1 receptor antagonist, CD96/PVR inhibitor, and mast cell stabilizer. This compound has high affinity for neuronal octapamine receptors in locusts (Ki = 2 nM) and honeybees (Ki = 1.1 nM). It is capable of inhibiting TARC, IL-8, and IL-4, activating anti-colon cancer immunity, and inhibiting Substance P-induced scratching behavior and increased vascular permeability. Epinastine hydrochloride is suitable for research into allergic diseases.
  • Selective and orally active histamine H1 receptor antagonist
  • CD96/PVR inhibitor and mast cell stabilizer
  • Exhibits high affinity for neuronal octapamine receptors
  • Inhibits TARC, IL-8, and IL-4
  • Activates anti-colon cancer immunity
  • Inhibits Substance P-induced scratching behavior and increased vascular permeability
  • Applicable for research into allergic diseases
Catalog No. 50-004-08738 Supplier Medchemexpress LLC Supplier No. HYB0640A500MG
Epinastine hydrochloride is a selective and orally active histamine H1 receptor antagonist, CD96/PVR inhibitor, and mast cell stabilizer. This compound has high affinity for neuronal octapamine receptors in locusts (Ki = 2 nM) and honeybees (Ki = 1.1 nM). It is capable of inhibiting TARC, IL-8, and IL-4, activating anti-colon cancer immunity, and inhibiting Substance P-induced scratching behavior and increased vascular permeability. Epinastine hydrochloride is suitable for research into allergic diseases.
  • Selective and orally active histamine H1 receptor antagonist
  • CD96/PVR inhibitor and mast cell stabilizer
  • Exhibits high affinity for neuronal octapamine receptors
  • Inhibits TARC, IL-8, and IL-4
  • Activates anti-colon cancer immunity
  • Inhibits Substance P-induced scratching behavior and increased vascular permeability
  • Applicable for research into allergic diseases