Learn More
Eurofins Discoverx (-)-Epinephrine (+)-bitartrate salt, 10 mg. (92-1284)
Catalog No. 501999801
Encompass
(-)-Epinephrine (+)-bitartrate is the sympathomimetic adrenal medulla hormone and agonist for the adrenergic receptors alpha and beta. Stimulates the heart, including systemic vasoconstriction, and causes GI relaxation and bronchial dilation. Used for asthma and cardiac failure, as well as to delay absorption of local anesthetics. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9801 Supplier Eurofins Discoverx Supplier No. 921284
(-)-Epinephrine (+)-bitartrate is the sympathomimetic adrenal medulla hormone and agonist for the adrenergic receptors alpha and beta. Stimulates the heart, including systemic vasoconstriction, and causes GI relaxation and bronchial dilation. Used for asthma and cardiac failure, as well as to delay absorption of local anesthetics. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.