Learn More
Apexbio Technology LLC Epinephrine Bitartrate 51-42-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063470
Encompass_Preferred
Epinephrine bitartrate the bitartrate salt form of endogenous epinephrine is a catecholamine neurotransmitter capable of simultaneously activating - and -adrenergic receptors Through receptor-mediated interactions epinephrine bitartrate modulates diverse physiological processes including vascular tone regulation bronchodilation cardiac function modulation and central nervous system stress responses This compound serves as an important pharmacological tool for in vitro and in vivo research studies investigating adrenergic receptor signaling pathways cardiovascular physiology respiratory system dynamics autonomic nervous system functions and related physiological or pathological conditions
Catalog No. 50-000-63470 Supplier Apexbio Technology LLC Supplier No. B13585.1
Epinephrine bitartrate the bitartrate salt form of endogenous epinephrine is a catecholamine neurotransmitter capable of simultaneously activating - and -adrenergic receptors Through receptor-mediated interactions epinephrine bitartrate modulates diverse physiological processes including vascular tone regulation bronchodilation cardiac function modulation and central nervous system stress responses This compound serves as an important pharmacological tool for in vitro and in vivo research studies investigating adrenergic receptor signaling pathways cardiovascular physiology respiratory system dynamics autonomic nervous system functions and related physiological or pathological conditions