Learn More
Alpco Ltd Epinephrine ELISA
SDP
Supplier:  Alpco Ltd 17EPIHUE01.1
Encompass
The Epinephrine ELISA is an enzyme immunoassay for the quantitative determination of Adrenaline (Epinephrine) in plasma and urine. For Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3491