Learn More
Apexbio Technology LLC Epinephrine HCl 55-31-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062064
Encompass_Preferred
Epinephrine hydrochloride is an adrenergic receptor agonist functioning as both a hormone and neurotransmitter Structurally epinephrine hydrochloride is the hydrochloride salt form of epinephrine facilitating receptor binding and signal transduction mediated by adrenergic receptor subtypes ( and ) Upon receptor activation it initiates downstream signaling pathways through G-protein-coupled mechanisms influencing intracellular second messengers such as cAMP calcium influx and protein kinase activity In biomedical research epinephrine hydrochloride serves as a pharmacological tool for investigating adrenergic receptor-mediated physiological responses including cardiac function modulation bronchodilation metabolic processes and vasoconstriction Its controlled application aids in studying sympathetic nervous system mechanisms cellular signaling cascades cardiovascular pharmacology and stress response frameworks
Catalog No. 50-000-62064 Supplier Apexbio Technology LLC Supplier No. B13275.1
Epinephrine hydrochloride is an adrenergic receptor agonist functioning as both a hormone and neurotransmitter Structurally epinephrine hydrochloride is the hydrochloride salt form of epinephrine facilitating receptor binding and signal transduction mediated by adrenergic receptor subtypes ( and ) Upon receptor activation it initiates downstream signaling pathways through G-protein-coupled mechanisms influencing intracellular second messengers such as cAMP calcium influx and protein kinase activity In biomedical research epinephrine hydrochloride serves as a pharmacological tool for investigating adrenergic receptor-mediated physiological responses including cardiac function modulation bronchodilation metabolic processes and vasoconstriction Its controlled application aids in studying sympathetic nervous system mechanisms cellular signaling cascades cardiovascular pharmacology and stress response frameworks