Learn More
Sigma Aldrich Fine Chemicals Biosciences (±)-Epinephrine hydrochloride, 329-63-5, MFCD00050562, 5 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences E46425G
Encompass
Empirical Formula (Hill Notation): C9H13NO3 · HCl, Molecular Weight: 219.67, storage temp: 2-8°C, Synonym: (±)-Adrenalin, 4-(1-Hydroxy-2-[methylamino]ethyl)-1,2-benzenediol hydrochloride.
Catalog No. NC0564698
 Restricted Item