Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epinephrine, USP, 97-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602424 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-424 100 g Amber Glass Bottle
18-602-423 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-424 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. EP130100GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EP130, 51-43-4, C9H13NO3

Spectrum™ Chemical Epinephrine, USP is also known as adrenaline and is a hormone and neurotransmitter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51-43-4
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.21
InChI Key UCTWMZQNUQWSLP-VIFPVBQESA-N
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

CAS Max % 100%
Ignition Residue Negligible
Loss on Drying 2%
Packaging Amber Glass Bottle
Quantity 100 g
UN Number UN2811
Specific Rotation -50.0° to -54.0°
Percent Purity 97 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.