Learn More
Epigentek Inc EpiNext NGS Barcode (Index) Set-12 (144 reactions)
SDP
Catalog No. 501250826
Encompass_Preferred
The EpiNext NGS Barcode (Index) Set-12 is designed to construct multiplex DNA/RNA libraries used for next-generation sequencing with the Illumina platform including GAIIx, HiSeq, MiSeq, and NextSeq. This product includes 12 DNA barcodes, each of which contain an index and flow cell binding sequence and can be attached to the sample insert during library preparation. Multiplexing several samples in a single flow cell can be achieved by pooling with the EpiNext NGS Barcodes. Pooling barcoded samples into a single flow cell significantly reduces hands-on time and provides robust data quality in NGS.
Catalog No. 50-125-0826 Supplier Epigentek Inc Supplier No. P1060144
The EpiNext NGS Barcode (Index) Set-12 is designed to construct multiplex DNA/RNA libraries used for next-generation sequencing with the Illumina platform including GAIIx, HiSeq, MiSeq, and NextSeq. This product includes 12 DNA barcodes, each of which contain an index and flow cell binding sequence and can be attached to the sample insert during library preparation. Multiplexing several samples in a single flow cell can be achieved by pooling with the EpiNext NGS Barcodes. Pooling barcoded samples into a single flow cell significantly reduces hands-on time and provides robust data quality in NGS.