Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epinine HCl, MP Biomedicals

Catalog No. ICN222653 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222653 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222653 Supplier MP Biomedicals, Inc Supplier No. 0522265380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62-32-8
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.67
MDL Number MFCD00035074
InChI Key JCDRZCWRRLKLTB-UHFFFAOYSA-N
Synonym n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride
PubChem CID 86470
IUPAC Name hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride
SMILES [H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1

Specifications

Quantity 100 mg
Formula Weight 203.67
Chemical Name or Material Epinine HCL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.