Learn More
Cayman Chemical -EpInphrIn-d3 10mg
SDP
Catalog No. 502943408
Encompass_Preferred
An internal standard for the quantification of (±)-epinephrine by GC- or LC-MS
Catalog No. 50-294-3408 Supplier Cayman Chemical Supplier No. 4043410 MG
An internal standard for the quantification of (±)-epinephrine by GC- or LC-MS