Learn More
Cayman Chemical (±)-Epinephrine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1189977-29-4, 25 mg
SDP
Catalog No. 502942578
Encompass_Preferred
An internal standard for the quantification of (±)-epinephrine by GC- or LC-MS
Catalog No. 50-294-2578 Supplier Cayman Chemical Supplier No. 4043425 MG
An internal standard for the quantification of (±)-epinephrine by GC- or LC-MS