Learn More
Cayman Chemical (±)-Epinephrine (hydrochloride), Purity: ≥95%, CAS: 329-63-5, 1 g
SDP
Catalog No. 502985573
Encompass_Preferred
A natural neurotransmitter that is activates adrenoceptors (Kis = 15, 735, and 3,970 nM for α1A-, β2-, and β1-adrenergic receptors, respectively); may induce either contraction or relaxation of vascular smooth muscle, depending on adrenoceptor subtype expression
Catalog No. 50-298-5573 Supplier Cayman Chemical Supplier No. 212451 G
A natural neurotransmitter that is activates adrenoceptors (Kis = 15, 735, and 3,970 nM for α1A-, β2-, and β1-adrenergic receptors, respectively); may induce either contraction or relaxation of vascular smooth muscle, depending on adrenoceptor subtype expression