Learn More
Cayman Chemical Epioxytetracycline, Purity: ≥95%, CAS: 14206-58-7, 10 mg
SDP
Catalog No. 502940900
Encompass_Preferred
A degradation product of oxytetracycline; has been found in swine manure compost and wastewater; considered a pollutant
Catalog No. 50-294-0900 Supplier Cayman Chemical Supplier No. 2798710 MG
A degradation product of oxytetracycline; has been found in swine manure compost and wastewater; considered a pollutant