Learn More
Cayman Chemical EpIpregnolon 50mg
SDP
Catalog No. 502948551
Encompass_Preferred
A neurosteroid and an active metabolite of pregnenolone; inhibits spontaneous contractions in myometrial strips isolated from at-term pregnant women (IC50 = 156 µM); decreases operant alcohol self-administration in rats at 10 and 20 mg/kg
Catalog No. 50-294-8551 Supplier Cayman Chemical Supplier No. 3429550 MG
A neurosteroid and an active metabolite of pregnenolone; inhibits spontaneous contractions in myometrial strips isolated from at-term pregnant women (IC50 = 156 µM); decreases operant alcohol self-administration in rats at 10 and 20 mg/kg